The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001172562.1 | NP_001166033.1 | 3 - 4 | 262 | 77 |
NM_018250.3 | NP_060720.2 | 3 - 4 | 496 | 77 | |
XM_006716356.2 | XP_006716419.1 | 3 - 4 | 293 | 77 | |
XM_006716357.3 | XP_006716420.1 | 3 - 4 | 293 | 77 | |
XM_011544574.1 | XP_011542876.1 | 2 - 3 | 194 | 77 | |
XM_011544575.2 | XP_011542877.1 | 4 - 5 | 714 | 77 | |
XM_011544576.1 | XP_011542878.1 | 3 - 4 | 293 | 77 | |
GenBank mRNA | AK001733.1 | - | 3 - 4 | 277 | 77 |
AK225072.1 | - | 3 - 4 | 229 | 77 | |
AK298468.1 | - | 3 - 4 | 262 | 77 | |
BC016687.1 | - | 3 - 4 | 228 | 77 | |
BC025267.1 | - | 3 - 4 | 246 | 77 | |
DC339488.1 | - | 3 - 4 | 497 | 77 |