The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145159.2 | NP_001138631.1 | 7 - 8 | 983 | 82 |
NM_001172562.1 | NP_001166033.1 | 8 - 9 | 812 | 82 | |
NM_018250.3 | NP_060720.2 | 8 - 9 | 1046 | 82 | |
NR_026826.2 | 6 - 7 | 855 | 82 | ||
XM_006716356.2 | XP_006716419.1 | 8 - 9 | 843 | 82 | |
XM_006716357.3 | XP_006716420.1 | 8 - 9 | 843 | 82 | |
XM_011544573.1 | XP_011542875.1 | 7 - 8 | 780 | 82 | |
XM_011544574.1 | XP_011542876.1 | 7 - 8 | 744 | 82 | |
XM_011544575.2 | XP_011542877.1 | 9 - 10 | 1264 | 82 | |
GenBank mRNA | AK001733.1 | - | 8 - 9 | 827 | 82 |
AK225072.1 | - | 8 - 9 | 779 | 82 | |
AK225530.1 | - | 8 - 9 | 843 | 82 | |
AK295977.1 | - | 4 - 5 | 343 | 82 | |
AK298468.1 | - | 8 - 9 | 812 | 82 | |
AK300593.1 | - | 7 - 8 | 743 | 82 | |
AK304525.1 | - | 6 - 7 | 588 | 82 | |
BC016687.1 | - | 8 - 9 | 778 | 82 | |
BC025267.1 | - | 8 - 9 | 796 | 82 |