The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003425.3 | NP_003416.1 | 4 - 5 | 826 | 82 |
XM_011527267.1 | XP_011525569.1 | 5 - 6 | 909 | 82 | |
XM_011527269.1 | XP_011525571.1 | 4 - 5 | 822 | 82 | |
XM_011527271.1 | XP_011525573.1 | 4 - 5 | 539 | 82 | |
XM_011527273.1 | XP_011525575.1 | 3 - 4 | 739 | 82 | |
XM_017027217.1 | XP_016882706.1 | 6 - 7 | 1028 | 82 | |
XM_017027218.1 | XP_016882707.1 | 6 - 7 | 1024 | 82 | |
XM_017027219.1 | XP_016882708.1 | 5 - 6 | 906 | 82 | |
XM_017027220.1 | XP_016882709.1 | 6 - 7 | 740 | 82 | |
XM_017027221.1 | XP_016882710.1 | 6 - 7 | 1042 | 82 | |
XM_017027222.1 | XP_016882711.1 | 5 - 6 | 945 | 82 | |
XM_017027223.1 | XP_016882712.1 | 6 - 7 | 769 | 82 | |
XM_017027224.1 | XP_016882713.1 | 4 - 5 | 844 | 82 | |
XM_017027225.1 | XP_016882714.1 | 5 - 6 | 657 | 82 | |
XM_017027226.1 | XP_016882715.1 | 5 - 6 | 653 | 82 | |
XM_017027227.1 | XP_016882716.1 | 3 - 4 | 451 | 82 | |
XM_017027228.1 | XP_016882717.1 | 4 - 5 | 826 | 82 | |
XM_017027229.1 | XP_016882718.1 | 5 - 6 | 945 | 82 | |
XM_017027230.1 | XP_016882719.1 | 4 - 5 | 822 | 82 | |
GenBank mRNA | AK127739.1 | - | 4 - 5 | 826 | 82 |
BC037575.1 | - | 1 - 2 | 93 | 82 |