The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003425.3 | NP_003416.1 | 7 - 8 | 1102 | 61 |
XM_011527267.1 | XP_011525569.1 | 8 - 9 | 1185 | 61 | |
XM_011527269.1 | XP_011525571.1 | 7 - 8 | 1098 | 61 | |
XM_011527271.1 | XP_011525573.1 | 7 - 8 | 815 | 61 | |
XM_011527273.1 | XP_011525575.1 | 6 - 7 | 1015 | 61 | |
XM_017027217.1 | XP_016882706.1 | 9 - 10 | 1304 | 61 | |
XM_017027218.1 | XP_016882707.1 | 9 - 10 | 1300 | 61 | |
XM_017027219.1 | XP_016882708.1 | 8 - 9 | 1182 | 61 | |
XM_017027220.1 | XP_016882709.1 | 9 - 10 | 1016 | 61 | |
XM_017027221.1 | XP_016882710.1 | 9 - 10 | 1318 | 61 | |
XM_017027222.1 | XP_016882711.1 | 8 - 9 | 1221 | 61 | |
XM_017027223.1 | XP_016882712.1 | 9 - 10 | 1045 | 61 | |
XM_017027224.1 | XP_016882713.1 | 7 - 8 | 1120 | 61 | |
XM_017027225.1 | XP_016882714.1 | 8 - 9 | 933 | 61 | |
XM_017027226.1 | XP_016882715.1 | 8 - 9 | 929 | 61 | |
XM_017027227.1 | XP_016882716.1 | 6 - 7 | 727 | 61 | |
GenBank mRNA | AK127739.1 | - | 7 - 8 | 1102 | 61 |
BC037575.1 | - | 4 - 5 | 369 | 61 | |
L75847.1 | - | 1 - 2 | 114 | 61 | |
M67509.1 | - | 1 - 2 | 114 | 61 |