Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FBXO27
Entrez Gene ID
126433
Gene Name
F-box protein 27
Gene Aliases
FBG5, Fbx27
Chromosome Location
Chr.19: 39007064 - 39032596 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178820.4 NP_849142.1 3 - 4 600 73
XM_017026290.1 XP_016881779.1 3 - 4 559 73
GenBank mRNA AF436061.1 - 3 - 4 543 73
BC014527.2 - 3 - 4 562 73
BC030060.1 - 3 - 4 529 73
BX377760.2 - 3 - 4 600 73

View Full Product Details