The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015541.2 | NP_056356.2 | 18 - 19 | 3073 | 112 |
XM_011533578.1 | XP_011531880.1 | 17 - 18 | 3530 | 112 | |
XM_011533579.2 | XP_011531881.1 | 18 - 19 | 2873 | 112 | |
XM_017006134.1 | XP_016861623.1 | 17 - 18 | 3599 | 112 | |
XM_017006135.1 | XP_016861624.1 | 13 - 14 | 2396 | 112 | |
XM_017006136.1 | XP_016861625.1 | 18 - 19 | 2937 | 112 | |
GenBank mRNA | AB050468.1 | - | 18 - 19 | 3581 | 112 |
AF381545.1 | - | 18 - 19 | 3073 | 112 | |
AK126745.1 | - | 11 - 12 | 2441 | 112 | |
AK294984.1 | - | 10 - 11 | 2075 | 112 | |
AK314103.1 | - | 18 - 19 | 3584 | 112 | |
AL117666.1 | - | 5 - 6 | 1230 | 112 | |
AY730707.1 | - | 17 - 18 | 3090 | 112 | |
BC014276.2 | - | 5 - 6 | 1032 | 112 | |
BC071561.1 | - | 19 - 20 | 3594 | 112 | |
BC140907.1 | - | 18 - 19 | 3079 | 112 |