Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TROAP
Entrez Gene ID
10024
Gene Name
trophinin associated protein
Gene Aliases
TASTIN
Chromosome Location
Chr.12: 49322135 - 49331733 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005480.3 NP_005471.3 12 - 13 1464 105
XM_006719181.1 XP_006719244.1 12 - 13 1408 105
XM_011537724.1 XP_011536026.1 11 - 12 1204 105
GenBank mRNA AK315211.1 - 12 - 13 1408 105
BC032583.1 - 12 - 13 1376 105
U04810.2 - 12 - 13 1407 105

View Full Product Details