Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TROAP
Entrez Gene ID
10024
Gene Name
trophinin associated protein
Gene Aliases
TASTIN
Chromosome Location
Chr.12: 49322135 - 49331733 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005480.3 NP_005471.3 9 - 10 1181 66
XM_006719181.1 XP_006719244.1 9 - 10 1125 66
XM_011537723.1 XP_011536025.1 9 - 10 1125 66
XM_011537724.1 XP_011536026.1 8 - 9 921 66
GenBank mRNA AK299302.1 - 9 - 10 988 66
AK309716.1 - 6 - 7 602 66
AK310709.1 - 8 - 9 919 66
AK315211.1 - 9 - 10 1125 66
BC032583.1 - 9 - 10 1093 66
U04810.2 - 9 - 10 1124 66

View Full Product Details