The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002062.4 | NP_002053.3 | 10 - 11 | 1104 | 98 |
NR_136562.1 | 10 - 11 | 1104 | 98 | ||
NR_136563.1 | 10 - 11 | 1104 | 98 | ||
XM_017010750.1 | XP_016866239.1 | 9 - 10 | 1639 | 98 | |
GenBank mRNA | BC112126.1 | - | 10 - 11 | 1116 | 98 |
BC113493.1 | - | 10 - 11 | 1104 | 98 | |
KR138540.1 | - | 10 - 11 | 1070 | 98 | |
L23503.1 | - | 10 - 11 | 1069 | 98 | |
U01104.1 | - | 10 - 11 | 1086 | 98 | |
U01156.1 | - | 10 - 11 | 1044 | 98 | |
U01157.1 | - | 10 - 11 | 1044 | 98 | |
U10037.1 | - | 10 - 11 | 1170 | 98 |