The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002062.4 | NP_002053.3 | 11 - 12 | 1242 | 81 |
NR_136562.1 | 11 - 12 | 1242 | 81 | ||
NR_136563.1 | 11 - 12 | 1242 | 81 | ||
XM_017010750.1 | XP_016866239.1 | 10 - 11 | 1777 | 81 | |
GenBank mRNA | BC112126.1 | - | 11 - 12 | 1254 | 81 |
BC113493.1 | - | 11 - 12 | 1242 | 81 | |
KR138540.1 | - | 11 - 12 | 1208 | 81 | |
L23503.1 | - | 11 - 12 | 1207 | 81 | |
U01104.1 | - | 11 - 12 | 1224 | 81 | |
U01156.1 | - | 11 - 12 | 1182 | 81 | |
U01157.1 | - | 11 - 12 | 1182 | 81 | |
U10037.1 | - | 11 - 12 | 1308 | 81 |