The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002062.4 | NP_002053.3 | 8 - 9 | 945 | 67 |
NR_136562.1 | 8 - 9 | 945 | 67 | ||
NR_136563.1 | 8 - 9 | 945 | 67 | ||
XM_017010750.1 | XP_016866239.1 | 7 - 8 | 1480 | 67 | |
GenBank mRNA | BC112126.1 | - | 8 - 9 | 957 | 67 |
BC113493.1 | - | 8 - 9 | 945 | 67 | |
KR138540.1 | - | 8 - 9 | 911 | 67 | |
L23503.1 | - | 8 - 9 | 910 | 67 | |
U01156.1 | - | 8 - 9 | 885 | 67 | |
U01157.1 | - | 8 - 9 | 885 | 67 | |
U10037.1 | - | 8 - 9 | 1011 | 67 |