The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042646.2 | NP_001036111.1 | 2 - 3 | 506 | 70 |
NM_001265608.1 | NP_001252537.1 | 2 - 3 | 506 | 70 | |
XM_005264956.2 | XP_005265013.1 | 2 - 3 | 314 | 70 | |
XM_005264960.3 | XP_005265017.1 | 2 - 3 | 314 | 70 | |
XM_005264962.3 | XP_005265019.1 | 2 - 3 | 314 | 70 | |
XM_005264963.3 | XP_005265020.1 | 2 - 3 | 314 | 70 | |
XM_006713029.2 | XP_006713092.1 | 2 - 3 | 314 | 70 | |
XM_006713030.2 | XP_006713093.1 | 2 - 3 | 314 | 70 | |
XM_006713031.3 | XP_006713094.1 | 2 - 3 | 314 | 70 | |
XM_011533489.2 | XP_011531791.1 | 2 - 3 | 314 | 70 | |
XM_017005906.1 | XP_016861395.1 | 4 - 5 | 927 | 70 | |
XM_017005909.1 | XP_016861398.1 | 3 - 4 | 812 | 70 | |
XM_017005911.1 | XP_016861400.1 | 3 - 4 | 812 | 70 | |
XM_017005912.1 | XP_016861401.1 | 2 - 3 | 314 | 70 | |
XM_017005913.1 | XP_016861402.1 | 2 - 3 | 314 | 70 | |
GenBank mRNA | AB028965.1 | - | 2 - 3 | 506 | 70 |
AK295480.1 | - | 2 - 3 | 544 | 70 | |
AK295848.1 | - | 2 - 3 | 413 | 70 | |
AK316260.1 | - | 2 - 3 | 403 | 70 |