Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KLHL13
Entrez Gene ID
90293
Gene Name
kelch like family member 13
Gene Aliases
BKLHD2
Chromosome Location
Chr.X: 117897813 - 118117340 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001168299.1 NP_001161771.1 5 - 6 710 78
NM_001168300.1 NP_001161772.1 4 - 5 958 78
NM_001168301.1 NP_001161773.1 5 - 6 608 78
NM_001168302.1 NP_001161774.1 5 - 6 902 78
NM_001168303.1 NP_001161775.1 4 - 5 828 78
NM_033495.3 NP_277030.2 4 - 5 1478 78
XM_011531409.2 XP_011529711.1 6 - 7 1370 78
XM_011531410.2 XP_011529712.1 6 - 7 670 78
XM_011531411.1 XP_011529713.1 5 - 6 1049 78
XM_017029950.1 XP_016885439.1 4 - 5 1739 78
GenBank mRNA AB037730.1 - 3 - 4 2837 78
AK122724.1 - 4 - 5 545 78
AK125356.1 - 4 - 5 1478 78
AK296324.1 - 5 - 6 617 78
AK297523.1 - 4 - 5 555 78
AK299257.1 - 4 - 5 722 78
AK302713.1 - 5 - 6 608 78
AK316509.1 - 5 - 6 862 78
AL591986.1 - 4 - 5 625 78
BC064576.1 - 4 - 5 630 78

View Full Product Details