Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
JAK3
Entrez Gene ID
3718
Gene Name
Janus kinase 3
Gene Aliases
JAK-3, JAK3_HUMAN, JAKL, L-JAK, LJAK
Chromosome Location
Chr.19: 17824782 - 17848071 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000215.3 NP_000206.2 14 - 15 2017 83
XM_005259896.3 XP_005259953.1 14 - 15 2056 83
XM_011527990.1 XP_011526292.1 14 - 15 2056 83
GenBank mRNA U09607.1 - 13 - 14 2012 83
U31601.1 - 14 - 15 2012 83

View Full Product Details