Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
JAK3
Entrez Gene ID
3718
Gene Name
Janus kinase 3
Gene Aliases
JAK-3, JAK3_HUMAN, JAKL, L-JAK, LJAK
Chromosome Location
Chr.19: 17824782 - 17848071 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000215.3 NP_000206.2 2 - 3 290 148
XM_005259896.3 XP_005259953.1 2 - 3 329 148
XM_011527990.1 XP_011526292.1 2 - 3 329 148
XM_011527991.2 XP_011526293.1 2 - 3 329 148
GenBank mRNA AK296383.1 - 2 - 3 291 148
BC028068.1 - 2 - 3 280 148
DA405504.1 - 2 - 3 291 148
KU178153.1 - 1 - 2 190 148
KU178154.1 - 1 - 2 190 148
U09607.1 - 1 - 2 285 148
U31601.1 - 2 - 3 285 148

View Full Product Details