The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002184.3 | NP_002175.2 | 10 - 11 | 1580 | 94 |
NM_175767.2 | NP_786943.1 | 9 - 10 | 1497 | 94 | |
NR_120480.1 | 11 - 12 | 1626 | 94 | ||
XM_017009441.1 | XP_016864930.1 | 10 - 11 | 1482 | 94 | |
XM_017009442.1 | XP_016864931.1 | 9 - 10 | 1459 | 94 | |
GenBank mRNA | AB015706.1 | - | 8 - 9 | 1276 | 94 |
BC117402.1 | - | 9 - 10 | 1379 | 94 | |
BC117404.1 | - | 9 - 10 | 1379 | 94 | |
HQ628629.1 | - | 8 - 9 | 1267 | 94 | |
M57230.1 | - | 10 - 11 | 1522 | 94 |