The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001190981.1 | NP_001177910.1 | 15 - 16 | 2154 | 122 |
NM_002184.3 | NP_002175.2 | 16 - 17 | 2337 | 122 | |
NM_175767.2 | NP_786943.1 | 15 - 16 | 2254 | 122 | |
NR_120480.1 | 17 - 18 | 2383 | 122 | ||
XM_017009441.1 | XP_016864930.1 | 16 - 17 | 2239 | 122 | |
XM_017009442.1 | XP_016864931.1 | 15 - 16 | 2216 | 122 | |
GenBank mRNA | AB102802.1 | - | 13 - 14 | 1841 | 122 |
BC117402.1 | - | 15 - 16 | 2136 | 122 | |
BC117404.1 | - | 15 - 16 | 2136 | 122 | |
HQ628629.1 | - | 13 - 14 | 1922 | 122 | |
HQ628630.1 | - | 13 - 14 | 1841 | 122 | |
M57230.1 | - | 16 - 17 | 2279 | 122 |