The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145255.2 | NP_001138727.1 | 4 - 5 | 927 | 70 |
GenBank mRNA | AK000593.1 | - | 4 - 5 | 632 | 70 |
AY429546.1 | - | 3 - 4 | 522 | 70 |