Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MIER3
Entrez Gene ID
166968
Gene Name
MIER family member 3
Gene Aliases
-
Chromosome Location
Chr.5: 56919602 - 56952130 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001297598.1 NP_001284527.1 9 - 10 869 67
NM_001297599.1 NP_001284528.1 9 - 10 854 67
XM_011543216.1 XP_011541518.1 9 - 10 889 67
XM_017009149.1 XP_016864638.1 10 - 11 890 67
GenBank mRNA AK093273.1 - 7 - 8 781 67
AK299285.1 - 8 - 9 829 67
BX537798.1 - 9 - 10 839 67
BX640907.1 - 9 - 10 857 67
CR749246.1 - 10 - 11 889 67
CR749805.1 - 10 - 11 889 67

View Full Product Details