The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020151.3 | NP_064536.2 | 3 - 4 | 950 | 72 |
XM_011511499.1 | XP_011509801.1 | 3 - 4 | 567 | 72 | |
GenBank mRNA | AF270647.1 | - | 3 - 4 | 877 | 72 |
AK023234.1 | - | 2 - 3 | 224 | 72 | |
AK054615.1 | - | 3 - 4 | 316 | 72 | |
AK291190.1 | - | 3 - 4 | 332 | 72 | |
AL542720.3 | - | 3 - 4 | 951 | 72 | |
BC003028.1 | - | 3 - 4 | 933 | 72 | |
BC007894.2 | - | 4 - 5 | 708 | 72 | |
BC008894.2 | - | 3 - 4 | 933 | 72 | |
BC009998.2 | - | 4 - 5 | 708 | 72 | |
BC012774.2 | - | 3 - 4 | 900 | 72 | |
BC012793.2 | - | 3 - 4 | 914 | 72 | |
BC013279.2 | - | 3 - 4 | 933 | 72 | |
BC014076.2 | - | 3 - 4 | 910 | 72 | |
BC014274.2 | - | 3 - 4 | 929 | 72 | |
BC032106.1 | - | 3 - 4 | 924 | 72 |