The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001010938.1 | NP_001010938.1 | 9 - 10 | 1592 | 71 |
NM_001308046.1 | NP_001294975.1 | 9 - 10 | 1650 | 71 | |
NM_005781.4 | NP_005772.3 | 9 - 10 | 1801 | 71 | |
XM_005269268.3 | XP_005269325.1 | 9 - 10 | 1592 | 71 | |
XM_005269270.3 | XP_005269327.1 | 9 - 10 | 1440 | 71 | |
XM_005269274.3 | XP_005269331.1 | 6 - 7 | 857 | 71 | |
XM_005269275.3 | XP_005269332.1 | 8 - 9 | 1336 | 71 | |
XM_011512317.2 | XP_011510619.1 | 11 - 12 | 2159 | 71 | |
XM_011512318.2 | XP_011510620.2 | 9 - 10 | 1531 | 71 | |
XM_011512320.1 | XP_011510622.1 | 9 - 10 | 1314 | 71 | |
XM_011512321.2 | XP_011510623.1 | 7 - 8 | 1131 | 71 | |
XM_017005508.1 | XP_016860997.1 | 9 - 10 | 1531 | 71 | |
XM_017005509.1 | XP_016860998.1 | 9 - 10 | 1531 | 71 | |
XM_017005510.1 | XP_016860999.1 | 9 - 10 | 1531 | 71 | |
GenBank mRNA | AB209338.1 | - | 6 - 7 | 3659 | 71 |
AK131539.1 | - | 9 - 10 | 1592 | 71 | |
AK225786.1 | - | 9 - 10 | 1597 | 71 | |
BC020389.1 | - | 8 - 9 | 1363 | 71 | |
BC028164.1 | - | 9 - 10 | 1315 | 71 | |
L13738.2 | - | 9 - 10 | 1876 | 71 |