The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031475.2 | NP_113663.2 | 1 - 2 | 460 | 64 |
XM_005263501.2 | XP_005263558.1 | 1 - 2 | 472 | 64 | |
XM_011542231.1 | XP_011540533.1 | 1 - 2 | 472 | 64 | |
XM_011542232.1 | XP_011540534.1 | 1 - 2 | 472 | 64 | |
XM_011542237.1 | XP_011540539.1 | 1 - 2 | 472 | 64 | |
XM_011542238.2 | XP_011540540.1 | 1 - 2 | 472 | 64 | |
XM_017002433.1 | XP_016857922.1 | 1 - 2 | 472 | 64 | |
XM_017002434.1 | XP_016857923.1 | 1 - 2 | 472 | 64 | |
GenBank mRNA | AL136880.1 | - | 1 - 2 | 460 | 64 |