The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_023925.4 | NP_076414.2 | 12 - 13 | 2899 | 82 |
XM_006719142.3 | XP_006719205.1 | 12 - 13 | 3022 | 82 | |
GenBank mRNA | AY074490.1 | - | 12 - 13 | 2899 | 82 |