The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005738.4 | NP_005729.1 | 1 - 2 | 441 | 122 |
GenBank mRNA | AK313233.1 | - | 1 - 2 | 94 | 122 |
BM550177.1 | - | 1 - 2 | 61 | 122 | |
BQ948724.1 | - | 1 - 2 | 608 | 124 | |
BU786406.1 | - | 1 - 2 | 76 | 122 | |
U73960.1 | - | 1 - 2 | 62 | 122 |