Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SNX10
Entrez Gene ID
29887
Gene Name
sorting nexin 10
Gene Aliases
OPTB8
Chromosome Location
Chr.7: 26291895 - 26374330 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001318198.1 NP_001305127.1 3 - 4 358 75
XM_017012085.1 XP_016867574.1 4 - 5 452 75
XM_017012086.1 XP_016867575.1 4 - 5 891 75
XM_017012087.1 XP_016867576.1 3 - 4 381 75
GenBank mRNA AK296142.1 - 3 - 4 188 75
AK316088.1 - 3 - 4 334 75

View Full Product Details