Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CHEK2
Entrez Gene ID
11200
Gene Name
checkpoint kinase 2
Gene Aliases
CDS1, CHK2, HuCds1, LFS2, PP1425, RAD53, hCds1
Chromosome Location
Chr.22: 28687743 - 28741866 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005735.1 NP_001005735.1 7 - 8 994 125
NM_001257387.1 NP_001244316.1 7 - 8 979 125
NM_007194.3 NP_009125.1 6 - 7 865 125
NM_145862.2 NP_665861.1 6 - 7 865 125
XM_006724114.3 XP_006724177.2 2 - 3 508 125
XM_006724116.2 XP_006724179.2 2 - 3 253 125
XM_011529839.2 XP_011528141.1 7 - 8 1016 125
XM_011529840.2 XP_011528142.1 7 - 8 1018 125
XM_011529841.1 XP_011528143.1 6 - 7 797 125
XM_011529842.2 XP_011528144.1 5 - 6 685 125
XM_011529843.1 XP_011528145.1 5 - 6 664 125
XM_011529844.2 XP_011528146.1 7 - 8 1026 125
XM_011529845.2 XP_011528147.1 5 - 6 572 125
XM_017028560.1 XP_016884049.1 7 - 8 980 125
XM_017028561.1 XP_016884050.1 3 - 4 224 125
GenBank mRNA AB040105.1 - 5 - 6 793 125
AF086904.1 - 5 - 6 793 125
AF096279.1 - 5 - 6 793 125
AF174135.1 - 5 - 6 793 125
AF217975.1 - 7 - 8 1578 125
AJ131197.1 - 5 - 6 793 125
AK289360.1 - 6 - 7 844 125
AK290754.1 - 6 - 7 867 125
AY551297.1 - 6 - 7 922 125
AY551299.1 - 5 - 6 793 125
AY551300.1 - 5 - 6 793 125
AY551301.1 - 5 - 6 683 125
AY551302.1 - 4 - 5 702 125
AY551303.1 - 3 - 4 520 125
AY551305.1 - 6 - 7 907 125
BC004207.2 - 6 - 7 820 125
CR456418.1 - 7 - 8 993 125

View Full Product Details