The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282112.1 | NP_001269041.1 | 6 - 7 | 792 | 97 |
NM_001282113.1 | NP_001269042.1 | 6 - 7 | 785 | 97 | |
NM_003935.4 | NP_003926.1 | 6 - 7 | 1108 | 97 | |
XM_005261811.1 | XP_005261868.1 | 8 - 9 | 958 | 97 | |
XM_005261813.2 | XP_005261870.1 | 7 - 8 | 823 | 97 | |
XM_006724349.1 | XP_006724412.1 | 7 - 8 | 769 | 97 | |
XM_006724350.1 | XP_006724413.1 | 7 - 8 | 943 | 97 | |
XM_011530482.1 | XP_011528784.1 | 7 - 8 | 1025 | 97 | |
XM_017029038.1 | XP_016884527.1 | 6 - 7 | 771 | 97 | |
XM_017029039.1 | XP_016884528.1 | 8 - 9 | 833 | 97 | |
XM_017029040.1 | XP_016884529.1 | 7 - 8 | 809 | 97 | |
XM_017029041.1 | XP_016884530.1 | 8 - 9 | 851 | 97 | |
GenBank mRNA | AF017146.1 | - | 5 - 6 | 693 | 97 |
AF053082.1 | - | 5 - 6 | 693 | 97 | |
AF125216.1 | - | 5 - 6 | 653 | 97 | |
AK290997.1 | - | 6 - 7 | 738 | 97 | |
AL833505.1 | - | 7 - 8 | 822 | 97 | |
BC002432.2 | - | 6 - 7 | 1015 | 97 | |
CR456596.1 | - | 5 - 6 | 607 | 97 |