Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CALB2
Entrez Gene ID
794
Gene Name
calbindin 2
Gene Aliases
CAB29, CAL2, CR
Chromosome Location
Chr.16: 71358713 - 71390438 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001740.4 NP_001731.2 5 - 6 478 74
NM_007088.3 NP_009019.1 5 - 6 478 74
NR_027910.2 5 - 6 478 74
GenBank mRNA AK222495.1 - 5 - 6 468 74
AK222648.1 - 5 - 6 468 74
AK291096.1 - 5 - 6 468 74
BC015484.2 - 5 - 6 453 74
DA060805.1 - 5 - 6 478 74
X56667.1 - 5 - 6 441 74

View Full Product Details