The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243077.1 | NP_001230006.1 | 1 - 2 | 279 | 70 |
NM_006139.3 | NP_006130.1 | 1 - 2 | 279 | 70 | |
XM_011512197.2 | XP_011510499.1 | 1 - 2 | 130 | 70 | |
GenBank mRNA | AF222341.1 | - | 1 - 2 | 57 | 70 |
AF222343.1 | - | 1 - 2 | 57 | 70 | |
AJ295273.1 | - | 1 - 2 | 57 | 70 | |
AK292986.1 | - | 1 - 2 | 61 | 70 | |
AK303421.1 | - | 1 - 2 | 116 | 70 | |
AK313313.1 | - | 1 - 2 | 203 | 70 | |
BC093698.1 | - | 1 - 2 | 97 | 70 | |
BC112085.1 | - | 1 - 2 | 97 | 70 | |
J02988.1 | - | 1 - 2 | 156 | 70 |