Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CD28
Entrez Gene ID
940
Gene Name
CD28 molecule
Gene Aliases
Tp44
Chromosome Location
Chr.2: 203706475 - 203739756 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011512195.2 XP_011510497.1 2 - 3 168 101
XM_011512197.2 XP_011510499.1 2 - 3 190 101
GenBank mRNA AF222343.1 - 2 - 3 117 101

View Full Product Details