Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CD28
Entrez Gene ID
940
Gene Name
CD28 molecule
Gene Aliases
Tp44
Chromosome Location
Chr.2: 203706475 - 203739756 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006139.3 NP_006130.1 2 - 3 630 84
XM_011512194.2 XP_011510496.1 2 - 3 465 84
XM_011512195.2 XP_011510497.1 3 - 4 207 84
XM_011512197.2 XP_011510499.1 3 - 4 229 84
GenBank mRNA AF222343.1 - 3 - 4 156 84
AJ517504.1 - 2 - 3 450 84
AK292986.1 - 2 - 3 412 84
AK313313.1 - 2 - 3 554 84
BC093698.1 - 2 - 3 448 84
BC112085.1 - 2 - 3 448 84
J02988.1 - 2 - 3 507 84

View Full Product Details