The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001195297.1 | NP_001182226.1 | 3 - 4 | 441 | 134 |
NM_138731.6 | NP_620059.1 | 2 - 3 | 302 | 134 | |
XM_017020992.1 | XP_016876481.1 | 4 - 5 | 551 | 134 | |
XM_017021009.1 | XP_016876498.1 | 2 - 3 | 283 | 134 | |
XM_017021012.1 | XP_016876501.1 | 2 - 3 | 283 | 134 | |
GenBank mRNA | AY059470.1 | - | 3 - 4 | 266 | 134 |