Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WASL
Entrez Gene ID
8976
Gene Name
Wiskott-Aldrich syndrome like
Gene Aliases
N-WASP, NWASP, WASPB
Chromosome Location
Chr.7: 123681927 - 123749071 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003941.3 NP_003932.3 6 - 7 963 135
GenBank mRNA AK289795.1 - 6 - 7 876 135
AM295156.1 - 6 - 7 626 135
BC052955.1 - 6 - 7 913 135
CR749438.1 - 7 - 8 868 135

View Full Product Details