Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OXA1L
Entrez Gene ID
5018
Gene Name
OXA1L, mitochondrial inner membrane protein
Gene Aliases
OXA1
Chromosome Location
Chr.14: 22766522 - 22771789 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005015.3 NP_005006.3 1 - 2 246 73
GenBank mRNA AK294849.1 - 1 - 2 93 73
AK298249.1 - 1 - 2 90 73
BC001669.2 - 1 - 2 72 73
BC112331.1 - 1 - 2 246 73
BP303816.1 - 1 - 2 78 73
BP330984.1 - 1 - 2 110 73
BX248295.1 - 1 - 2 65 73
X80695.1 - 1 - 2 72 73

View Full Product Details