Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANKRD13A
Entrez Gene ID
88455
Gene Name
ankyrin repeat domain 13A
Gene Aliases
ANKRD13, NY-REN-25
Chromosome Location
Chr.12: 109999169 - 110039432 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_017020157.1 XP_016875646.1 11 - 12 1495 157
XM_017020158.1 XP_016875647.1 11 - 12 1489 157
GenBank mRNA AF155103.1 - 4 - 5 434 157

View Full Product Details