Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KCTD10
Entrez Gene ID
83892
Gene Name
potassium channel tetramerization domain containing 10
Gene Aliases
BTBD28, MSTP028, ULRO61, hBACURD3
Chromosome Location
Chr.12: 109448655 - 109477544 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001317399.1 NP_001304328.1 3 - 4 662 76
NM_031954.4 NP_114160.1 3 - 4 662 76
NR_133898.1 2 - 3 448 76
GenBank mRNA AF113208.1 - 3 - 4 407 76
AK027543.1 - 3 - 4 468 76
AK222548.1 - 3 - 4 418 76
AK309783.1 - 2 - 3 277 76
AY205299.1 - 3 - 4 468 76
AY597809.1 - 2 - 3 380 76
BC040062.1 - 3 - 4 415 76
CX785497.1 - 2 - 3 191 76

View Full Product Details