The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004385.4 | NP_004376.2 | 7 - 8 | 4354 | 153 |
GenBank mRNA | AB209491.1 | - | 7 - 8 | 4237 | 153 |
U16306.1 | - | 7 - 8 | 4264 | 153 |