The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001306212.1 | NP_001293141.1 | 22 - 23 | 2904 | 71 |
NM_001306213.1 | NP_001293142.1 | 22 - 23 | 2956 | 71 | |
NM_001306214.1 | NP_001293143.1 | 21 - 22 | 2898 | 71 | |
NM_003248.5 | NP_003239.2 | 21 - 22 | 3070 | 71 | |
GenBank mRNA | AK302842.1 | - | 21 - 22 | 2898 | 71 |
AK313587.1 | - | 21 - 22 | 2886 | 71 | |
BC050456.1 | - | 22 - 23 | 3154 | 71 | |
Z19585.1 | - | 21 - 22 | 2858 | 71 |