The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001307951.1 | NP_001294880.1 | 1 - 2 | 248 | 67 |
XM_006718750.1 | XP_006718813.1 | 1 - 2 | 248 | 67 | |
GenBank mRNA | AK299264.1 | - | 1 - 2 | 229 | 67 |
DA363664.1 | - | 1 - 2 | 249 | 67 |