The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006473.3 | NP_006464.1 | 2 - 3 | 352 | 63 |
XM_005273714.1 | XP_005273771.1 | 2 - 3 | 244 | 63 | |
GenBank mRNA | AF069735.1 | - | 1 - 2 | 151 | 63 |
AK289499.1 | - | 2 - 3 | 262 | 63 | |
AK301145.1 | - | 2 - 3 | 252 | 63 | |
AK310054.1 | - | 2 - 3 | 253 | 63 | |
AK314330.1 | - | 2 - 3 | 238 | 63 | |
BC008785.2 | - | 2 - 3 | 252 | 63 | |
DC311213.1 | - | 2 - 3 | 352 | 63 |