The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014023.3 | NP_054742.2 | 13 - 14 | 1523 | 98 |
GenBank mRNA | AB023199.1 | - | 13 - 14 | 1494 | 98 |
AK128194.1 | - | 12 - 13 | 4309 | 98 | |
AK292591.1 | - | 13 - 14 | 1489 | 98 | |
AK307259.1 | - | 12 - 13 | 1416 | 98 | |
AL136827.1 | - | 13 - 14 | 1501 | 98 |