The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152227.2 | NP_689413.1 | 1 - 2 | 291 | 100 |
GenBank mRNA | AF121855.1 | - | 1 - 2 | 147 | 100 |
DA567854.1 | - | 1 - 2 | 291 | 100 |