Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
DA267627.1 |
- |
1 |
|
|
Chr. 20 - 2652171 - 2652873 [+] on Build GRCh38 |
105 |
GenBankmRNA |
AK021907.1 |
- |
1 |
732 |
|
Chr. 20 - 2650935 - 2652978 [-] on Build GRCh38 |
104 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
microRNA 1292 |
MIR1292 |
MIRN1292, hsa-mir-1292, mir-1292 |
|
Anti-sense |
Chr. - 2652777 - 2652842 on Build GRCh38 |
Downstream |
small nucleolar RNA, C/D box 110 |
SNORD110 |
HBII-55 |
1.2kb
|
Anti-sense |
Chr. - 2654212 - 2654286 on Build GRCh38 |
View Full Product Details