The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005564.4 | NP_005555.2 | 5 - 6 | 665 | 61 |
GenBank mRNA | AW778875.1 | - | 2 - 3 | 61 | |
BC033089.1 | - | 5 - 6 | 629 | 61 | |
BM977724.1 | - | 2 - 3 | 61 | ||
CA454137.1 | - | 5 - 6 | 627 | 61 | |
N79823.1 | - | 2 - 3 | 61 |