Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NEPRO
Entrez Gene ID
25871
Gene Name
nucleolus and neural progenitor protein
Gene Aliases
C3orf17, NET17
Chromosome Location
Chr.3: 113002444 - 113019733 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001319109.1 NP_001306038.1 6 - 7 1104 131
NM_001319110.1 NP_001306039.1 5 - 6 949 131
NM_001319111.1 NP_001306040.1 7 - 8 918 131
NM_001319112.1 NP_001306041.1 7 - 8 1117 131
NM_001319114.1 NP_001306043.1 7 - 8 1163 131
NM_001319115.1 NP_001306044.1 7 - 8 1159 131
NM_015412.3 NP_056227.2 8 - 9 1289 131
NR_134969.1 5 - 6 1042 131
XM_005247276.2 XP_005247333.1 8 - 9 1256 131
XM_011512635.2 XP_011510937.1 8 - 9 1011 131
XM_011512636.2 XP_011510938.1 8 - 9 1015 131
XM_011512638.2 XP_011510940.1 7 - 8 910 131
GenBank mRNA AK024325.1 - 8 - 9 1247 131
AK293218.1 - 5 - 6 980 131
AK295165.1 - 7 - 8 1048 131
AK295954.1 - 5 - 6 886 131
AK300811.1 - 7 - 8 851 131
AK301161.1 - 7 - 8 1077 131
AK304022.1 - 2 - 3 377 131
AL117573.1 - 8 - 9 1231 131
BC014673.2 - 7 - 8 1747 131
BC067743.1 - 8 - 9 1249 131
CR749341.1 - 6 - 7 1105 131

View Full Product Details