Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CEP97
Entrez Gene ID
79598
Gene Name
centrosomal protein 97
Gene Aliases
2810403B08Rik, LRRIQ2
Chromosome Location
Chr.3: 101724098 - 101770562 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001303401.1 NP_001290330.1 3 - 4 430 142
NM_024548.3 NP_078824.2 3 - 4 430 142
GenBank mRNA AK294784.1 - 3 - 4 368 142
AL833269.1 - 3 - 4 380 142
BC041085.1 - 3 - 4 373 142
DA520309.1 - 3 - 4 432 142

View Full Product Details