Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CEP97
Entrez Gene ID
79598
Gene Name
centrosomal protein 97
Gene Aliases
2810403B08Rik, LRRIQ2
Chromosome Location
Chr.3: 101724098 - 101770562 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001303401.1 NP_001290330.1 7 - 8 984 88
NM_024548.3 NP_078824.2 7 - 8 984 88
XM_006713743.3 XP_006713806.1 6 - 7 1375 88
XM_011513127.1 XP_011511429.1 7 - 8 941 88
XM_017007178.1 XP_016862667.1 6 - 7 1377 88
GenBank mRNA AK294784.1 - 7 - 8 922 88
AK309806.1 - 6 - 7 883 88
AL833269.1 - 7 - 8 934 88
BC041085.1 - 7 - 8 927 88

View Full Product Details