Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EBF3
Entrez Gene ID
253738
Gene Name
early B-cell factor 3
Gene Aliases
COE3, EBF-3, O/E-2, OE-2
Chromosome Location
Chr.10: 129835232 - 129964274 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005463.2 NP_001005463.1 6 - 7 604 81
XM_005252667.3 XP_005252724.1 6 - 7 1051 81
XM_005252668.3 XP_005252725.1 6 - 7 1051 81
XM_005252669.3 XP_005252726.1 6 - 7 1051 81
XM_006717739.3 XP_006717802.1 6 - 7 1051 81
XM_006717740.3 XP_006717803.1 6 - 7 1051 81
XM_006717741.3 XP_006717804.1 6 - 7 1051 81
XM_006717742.3 XP_006717805.1 6 - 7 1051 81
XM_006717743.3 XP_006717806.1 6 - 7 1051 81
XM_006717744.3 XP_006717807.1 6 - 7 1051 81
XM_011539574.2 XP_011537876.1 6 - 7 1051 81
XM_017016027.1 XP_016871516.1 6 - 7 1051 81
GenBank mRNA AL832828.1 - 6 - 7 531 81
BC126130.1 - 6 - 7 595 81
BC130479.1 - 6 - 7 595 81

View Full Product Details