Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WDR11
Entrez Gene ID
55717
Gene Name
WD repeat domain 11
Gene Aliases
BRWD2, DR11, HH14, SRI1, WDR15
Chromosome Location
Chr.10: 120851175 - 120909526 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018117.11 NP_060587.8 16 - 17 2365 69
XM_005269962.2 XP_005270019.1 16 - 17 2082 69
XM_005269963.2 XP_005270020.1 16 - 17 2214 69
XM_005269964.2 XP_005270021.1 9 - 10 1120 69
XM_017016397.1 XP_016871886.1 15 - 16 2053 69
XM_017016398.1 XP_016871887.1 15 - 16 1983 69
XM_017016399.1 XP_016871888.1 15 - 16 1927 69
XM_017016400.1 XP_016871889.1 16 - 17 2256 69
GenBank mRNA AB037772.2 - 16 - 17 2178 69
AF320223.1 - 16 - 17 2152 69
AK000579.1 - 2 - 3 167 69
AK001368.1 - 11 - 12 1268 69
AK299524.1 - 9 - 10 985 69
AK315145.1 - 16 - 17 2143 69
AL137699.1 - 14 - 15 1838 69
BC040469.1 - 16 - 17 2182 69
BC071564.1 - 16 - 17 2148 69

View Full Product Details