Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WDR11
Entrez Gene ID
55717
Gene Name
WD repeat domain 11
Gene Aliases
BRWD2, DR11, HH14, SRI1, WDR15
Chromosome Location
Chr.10: 120851175 - 120909526 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018117.11 NP_060587.8 20 - 21 2867 142
XM_005269962.2 XP_005270019.1 20 - 21 2584 142
XM_005269963.2 XP_005270020.1 20 - 21 2716 142
XM_005269964.2 XP_005270021.1 13 - 14 1622 142
XM_017016397.1 XP_016871886.1 19 - 20 2555 142
XM_017016398.1 XP_016871887.1 19 - 20 2485 142
XM_017016399.1 XP_016871888.1 19 - 20 2429 142
GenBank mRNA AB037772.2 - 20 - 21 2680 142
AF320223.1 - 20 - 21 2654 142
AK000579.1 - 6 - 7 669 142
AK001368.1 - 15 - 16 1770 142
AK025050.1 - 3 - 4 378 142
AK124522.1 - 7 - 8 3054 142
AK299524.1 - 13 - 14 1487 142
AK315145.1 - 20 - 21 2645 142
AL137699.1 - 18 - 19 2340 142
BC040469.1 - 20 - 21 2684 142
BC071564.1 - 20 - 21 2650 142

View Full Product Details